Skip to Content
Merck
All Photos(1)

Key Documents

256234

Sigma-Aldrich

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$99.80

$99.80


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$99.80

About This Item

Empirical Formula (Hill Notation):
C13H16ClNOS
CAS Number:
Molecular Weight:
269.79
Beilstein:
3788921
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$99.80


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

142-144 °C (lit.)

SMILES string

[Cl-].Cc1c(CCO)sc[n+]1Cc2ccccc2

InChI

1S/C13H16NOS.ClH/c1-11-13(7-8-15)16-10-14(11)9-12-5-3-2-4-6-12;/h2-6,10,15H,7-9H2,1H3;1H/q+1;/p-1

InChI key

IWSVLBKHBJGMAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service