Skip to Content
Merck
All Photos(1)

Key Documents

Y0001758

Thiamine nitrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Vitamin B1 nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C12H17N5O4S
CAS Number:
Molecular Weight:
327.36
Beilstein:
3857790
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

thiamine

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+]([O-])=O.CC1=NC=C(C[N+]2=CSC(CCO)=C2C)C(N)=N1

InChI

1S/C12H17N4OS.NO3/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;2-1(3)4/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);/q+1;-1

InChI key

UIERGBJEBXXIGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Thiamine nitrate is a mononitrate salt of thiamine. It is synthesized from thiamine hydrochloride following a sequence of steps. Thiamine nitrate is used as a dietary supplement in crackers, eggs substitute and enriched flour.

Application

Thiamine nitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service