Skip to Content
Merck
All Photos(1)

Documents

816104

Endfitting w/ Fixed 10 μm Pyrex® Frit for 5 mm ID Glass TSKgel® Columns

Pyrex frit, pkg of 1 ea

Synonym(s):

TSKgel® Hardware and Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

Pyrex frit
fitting (fluorocarbon)

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 16104

technique(s)

LPLC: suitable

pore size

10 μm

compatibility

Tosoh 5 mm ID TSKgel Glass Columns

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

PYREX is a registered trademark of Corning, Inc.
TSKgel is a registered trademark of Tosoh Corporation

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service