Skip to Content
Merck
All Photos(1)

Key Documents

805748

Endfitting with fixed 10 μm frit for all 7.5 mm I.D. TSKgel® stainless steel columns

stainless steel, pkg of 1 ea

Synonym(s):

TSKgel® Hardware and Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
pore size:
10 μm
Product 805748 is not currently sold in your country. Contact Technical Service

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 05748

technique(s)

HPLC: suitable

pore size

10 μm

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service