Skip to Content
Merck
All Photos(1)

Key Documents

813998

Endfitting with fixed 1 μm frit

stainless steel, pore size 1 μm, pkg of 1 ea

Synonym(s):

TSKgel® Hardware and Accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Product 813998 is not currently sold in your country. Contact Technical Service

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 13998

technique(s)

HPLC: suitable

pore size

1 μm

compatibility

Tosoh TSKgel® NPR columns

Looking for similar products? Visit Product Comparison Guide

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service