Skip to Content
Merck
All Photos(1)

Key Documents

59025-U

Union, reducing

for connecting 1/16 to 1/32 in., bore size 0.25 mm

Synonym(s):

Valco® Unions

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$75.40

$75.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$75.40

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$75.40


Check Cart for Availability

Request a Bulk Order

material

stainless steel union and fittings (nuts and ferrules included)

description

internal

packaging

pkg of 1 unit

bore size

0.25 mm

fitting

10-32 male Valco for 1/16 in. O.D. tubing
6-32 Valco for 1/32 in. O.D. tubing

compatibility

for connecting 1/16 to 1/32 in.

Looking for similar products? Visit Product Comparison Guide

General description

Stainless steel, unless noted otherwise. Nuts and ferrules included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service