Skip to Content
Merck
All Photos(1)

Key Documents

575654-U

Supelco

Disposable Reservoir / Waste Tray

configured for collecing eluate waste during PlatePrep SPE processing, PVC, pk of 25

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$301.00

$301.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
25 EA
$301.00

About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

$301.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

material

PVC

packaging

pk of 25

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

configured for collecing eluate waste during PlatePrep SPE processing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service