Key Documents
57358-U
Supelco
Vial puck, 15 mL, for SPME Sampling Stand
made to hold 8 × 15 mL vials
Select a Size
1 EA
$414.00
$414.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$414.00
About This Item
UNSPSC Code:
41115854
NACRES:
NB.21
$414.00
Check Cart for Availability
Request a Bulk Order
technique(s)
solid phase microextraction (SPME): suitable
application(s)
food and beverages
compatibility
made to hold 8 × 15 mL vials
Choose from one of the most recent versions:
Certificates of Analysis (COA)