Skip to Content
Merck
All Photos(1)

Documents

57228-U

Supelco

Discovery® DSC-WCX SPE Bulk Packing

pkg of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

packaging

pkg of 100 g

technique(s)

solid phase extraction (SPE): suitable

matrix active group

carboxypropyl bonding

application(s)

food and beverages

separation technique

ion exchange

General description

Retention Mechanism: Cation exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • A polymerically bonded, ethylenediamine triacetic acid phase with a pKa of 4.8
  • Counter ion is Na+
  • Ion exchange capacity is ∼ 0.15 meq/g
  • Carries a negative charge at pH 6.8 or above
  • A pH of 2.8 or below neutralizes this phase for easier elution of strong cationic analytes that are neutralized only at extreme basic conditions
  • Typically used when dealing with very strong cationic (high pKa) compounds that may be irreversibly retained on strong cation exchangers
·

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service