Skip to Content
Merck
All Photos(1)

Key Documents

54774

Supelco

LiChrospher® RP-18 (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,370.00

$1,370.00


Check Cart for Availability


Select a Size

Change View
1 EA
$1,370.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

$1,370.00


Check Cart for Availability

Product Name

LiChrospher® RP-18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Agency

suitable for USP L1

feature

endcapped: no

manufacturer/tradename

LiChrospher®

extent of labeling

21% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

350 m2/g

matrix

octadecyl
silica particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

LiChrospher® is a reliable and versatile spherical silica sorbent, which is traditionally produced and available with different modifications. LiChrospher® RP-18 (5 μm particle size, L × I.D. 15 cm × 4.6 mm) delivers highly reproducible results and is designed for reversed-phase chromatography of acidic, neutral and weakly basic compounds.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the pH range for LiChrospher 100 RP-18, 5 µm, 4.0 x 250 mm cartridge (AC)?

    1 answer
    1. The LiChrospher phase has a pH stability range of 2-7.6.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service