Skip to Content
Merck
All Photos(1)

Documents

54951

Supelco

LiChrosorb® RP-18 (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

LiChrosorb® RP-18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Agency

suitable for USP L1

feature

endcapped: no

manufacturer/tradename

LiChrosorb®

extent of labeling

17% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

300 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

LiChrosorb® RP-18 HPLC Columns are packed with the finest commercially available silicas and bonded phases from the most recognized HPLC manufacturers in the world. The columns are tested with specially selected solutes that can be used to establish the reproducibility of retention and selectivity needed in the HPLC applications. They are suitable for reversed-phase HPLC of neutral, acidic and weak basic compounds, according to L1 USP specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrosorb is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service