Skip to Content
Merck
All Photos(1)

Key Documents

50137-U

Supelco

LiChrospher® RP-18 (5 µm) HPLC Columns

L × I.D. 25 cm × 4 mm, HPLC Column

Synonym(s):

TM=["LiChrospher"] C18 HPLC column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,510.00

$1,510.00


Check Cart for Availability


Select a Size

Change View
1 EA
$1,510.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$1,510.00


Check Cart for Availability

Product Name

LiChrospher® RP-18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

LiChrospher®

feature

endcapped: no

manufacturer/tradename

LiChrospher®

packaging

1 ea of

extent of labeling

21% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4 mm

surface area

350 m2/g

matrix

octadecyl
silica particle platform
fully porous particle

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

operating pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

LiChrospher® RP-18 HPLC column (L × I.D. 25 cm × 4 mm) is found to be reliable and high versatile. Suitable for reversed-phase high performance liquid chromatography (RP-HPLC) of acidic, neutral and weakly basic compounds, according to L1 USP specifications.

Application


  • HPLC-UV and GC-MS Methods for Determination of Chlorambucil and Valproic Acid in Plasma for Further Exploring a New Combined Therapy of Chronic Lymphocytic Leukemia.: This study developed and validated HPLC-UV and GC-MS methods for determining chlorambucil and valproic acid in plasma, aiming to explore their combined therapy for chronic lymphocytic leukemia. The LiChrospher® RP-18 HPLC Column was instrumental in achieving the separation and quantification (Lipska et al., 2021).

  • Rapid Determination of Metronidazole and 2-Hydroxymetronidazole in Murine Blood Plasma.: This research utilized the LiChrospher® RP-18 HPLC Column for the rapid determination of metronidazole and its metabolite in murine blood plasma, highlighting its effectiveness in pharmacokinetic studies (Zemanová et al., 2022).

  • Application of chemometric approach for development and validation of high performance liquid chromatography method for estimation of ropinirole hydrochloride.: This paper presents a chemometric approach to developing and validating an HPLC method for estimating ropinirole hydrochloride, utilizing the LiChrospher® RP-18 column for precise and accurate results (Fatima et al., 2019).

  • A New Solid-Phase Extraction Method for Determination of Pantoprazole in Human Plasma Using High-Performance Liquid Chromatography.: The study developed a novel solid-phase extraction method for determining pantoprazole in human plasma using HPLC with the LiChrospher® RP-18 column, achieving high sensitivity and specificity (Zendelovska et al., 2019).

  • Application of an HPLC-MS/MS method for Teicoplanin drug substance and related impurities, part 2: Identity assignment of related impurities.: This research employed the LiChrospher® RP-18 column in an HPLC-MS/MS method to identify and assign related impurities in teicoplanin drug substances, demonstrating the column′s capability in impurity profiling (Tengattini et al., 2019).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service