Skip to Content
Merck
All Photos(1)

Key Documents

509825

SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Fixed Needle

250F-LC, volume 0.25 mL (250 μL), needle size 22 ga, needle L × O.D. 2 in. × 0.7112 mm (0.028 in.)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$120.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$120.00

About This Item

UNSPSC Code:
41122001

$120.00


Check Cart for Availability

Request a Bulk Order

material

blunt tip stainless steel needle (point style 3)

needle size

22 ga

feature

Fixed needle

packaging

pkg of 1 ea

manufacturer/tradename

SGE 006300

needle L × O.D.

2 in. × 0.7112 mm (0.028 in.)

volume

0.25 mL (250 μL)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC
Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service