Skip to Content
Merck
All Photos(1)

Key Documents

26248

SGE MicroVolume syringes with standard plunger, fixed needle

25F, volume 25 μL, needle size 25 ga, needle L 50 mm

Synonym(s):

SYRINGE SGE 25UL FN PT#2, 25GA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$88.50

$88.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$88.50

About This Item

UNSPSC Code:
41122001

$88.50


Check Cart for Availability

Request a Bulk Order

material

bevel tip stainless steel needle (point style 2)

needle size

25 ga

feature

needle type Fixed

packaging

pkg of 1 ea

manufacturer/tradename

SGE 003000

needle L

50 mm

volume

25 μL

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of solid-phase microextraction to the headspace gas chromatographic analysis of halogenated volatiles in selected foods.
Page, B. Denis, and Gladys Lacroix.
Journal of Chromatography A, 648.1, 199-211 (1993)
Application of Fourier transform infrared spectroscopy for monitoring short-chain free fatty acids in Swiss cheese.
Koca, N., et al.
Journal of Dairy Science, 90.8, 3596-3603 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service