Skip to Content
Merck
All Photos(1)

Key Documents

509817

SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Fixed Needle

100F-LC, volume 100 μL, needle size 22 ga, needle L × O.D. 2 in. × 0.028 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$91.20

$91.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$91.20

About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500

$91.20


Check Cart for Availability

Request a Bulk Order

material

blunt tip stainless steel needle (point style 3)

needle size

22 ga

feature

Fixed needle

packaging

pkg of 1 ea

manufacturer/tradename

SGE 005300

needle L × O.D.

2 in. × 0.028 in.

volume

100 μL

Looking for similar products? Visit Product Comparison Guide

General description

Designers of syringes for microliter sample injections face two interesting problems: extremely fine plungers and proportionally large sample hold-up in the needle. SGE′s Plunger-in-Needle feature, which permits extremely small sample injections, and the SuperFlex plunger, which resists bending and kinking, addresses both problems. Other features of the SGE Microvolume Syringes [100F-LC, volume 100 μL, needle size 22 ga (blunt tip), needle L × O.D. 2 in. × 0.028 in.] also include:
  • Sharp and clear fused graduation.
  • Extended graduation lines and wide white backing for reduced errors.
  • Angled needle tip for optimum septum penetration.
  • Standard plunger is stainless steel individually fitted to its barrel.
  • Plunger Protection – An option on some SGE microvolume syringes.
  • Extended protection tube from the top of the flange helps prevent plunger bending during injection.
  • Rugged Guided Plunger – An option on some SGE microvolume syringes.
  • Extended barrel guides the plunger during injection and is suitable for training and student use.
  • Robust

SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Fixed Needle is equipped with stainless steel needle blunt tip (point style #3). The 90° blunt tip has chamfered and polished edges that eliminate damage to the valve′s rotor seal and stator face. This style also can be used for pipetting of liquids.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC
Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service