Key Documents
509639
Syringes Fitted with Luer Lock Valve (509353)
volume 25 mL, 25MDR-VLLMA-GT, needle size (not included)
Select a Size
1 EA
$546.00
$546.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$546.00
About This Item
UNSPSC Code:
41122001
$546.00
Check Cart for Availability
Request a Bulk Order
material
glass syringe
needle size
(not included)
feature
Pre-fitted with Luer Lock Valve
packaging
pkg of 1 ea
manufacturer/tradename
SGE 009472
volume
25 mL
General description
<UL><LI>SGE Syringes Fitted with Luer Lock Valve [volume 25 mL, needle size (not included)], Model: 25MDR-VLLMA-GT.</LI><LI>Composed of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.</LI><LI>Autoclavable syringes but the plunger removal required.</LI><LI>Feature an interference fit between the plunger tip and the barrel that enhances the gas sealing ability of the syringe.
  • Maximum inertness for liquid or gas samples.
  • Suitable for manual dispenser/diluter applications.
  • High accuracy of dispensed volume.
  • A wide variety of terminations.
Choose from one of the most recent versions:
Certificates of Analysis (COA)