Skip to Content
Merck
All Photos(1)

Key Documents

509507

SGE Gas Tight Syringes, Luer Lock

10MDR-LL-GT, volume 10 mL, needle size (not included)

Synonym(s):

SYRINGE SGE 10ML, LUER LOCK

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$219.00

$219.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$219.00

About This Item

UNSPSC Code:
41122001

$219.00


Available to ship on16 April 2025Details


Request a Bulk Order

material

glass syringe

needle size

(not included)

feature

needle type Luer lock

packaging

pkg of 1 ea

manufacturer/tradename

SGE 008960

volume

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

Gas-tight syringes with Luer-lock push-button dispenser valves (SGE) are basically fitted to Luer-lock needles, and are easily removable.[1]

Application

SGE gas tight syringes may be used for introduction of samples to the gas sampling and injection system (GASIS) inlet for the analyses of compound-specific δD and δ13C gas mixtures using gas chromatography-oxidation/reduction-isotope ratio mass spectrometry.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Henning et al.
Rapid communications in mass spectrometry : RCM, 21(14), 2269-2272 (2007-06-20)
Compound-specific deltaD and delta13C analyses of gas mixtures are useful indicators of geochemical and environmental factors. However, the relative concentrations of individual components in gas mixtures (e.g., H2, CO2, methane, ethane, propane, i-butane, n-butane) may vary over several orders of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service