Skip to Content
Merck
All Photos(1)

Key Documents

503185

Supelco

Carbon Dioxide Trap

volume 100 cc, 1/8 in. fitting

Synonym(s):

CO2 TRAP 100CC, 1/8IN FITTINGS

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$437.00

$437.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$437.00

About This Item

EC Number:
UNSPSC Code:
12352200

$437.00


Check Cart for Availability

Request a Bulk Order

volume

100 cc

fitting

1/8 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide nonfibrous silicate material is universally accepted for its high absorptive capacity and indicating properties for carbon dioxide. Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required. As carbon dioxide is absorbed, the greenish-brown material turns white because of the formation of sodium carbonate. The trap body is constructed of borosilicate glass, and the fittings are nickel-plated and have sintered stainless steel frits.

A moisture trap should be installed downstream from this unit to absorb the moisture that is produced.

Note: Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent.

Dimensions:
  • 100 cc Traps: 123/4 in. (32.4 cm) long x 13/4 in. (44 mm) diameter
  • 250 cc Traps: 171/2 in. (44.4 cm) long x 2 in. (51 mm) diameter

Application

It may be used to connect combustion oven and GCMS separator, for transfer of the gas into MS inlet system which monitors isotope-ratio.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotope-ratio-monitoring gas chromatography-mass spectrometry.
Matthews, D. E., and J. M. Hayes.
Analytical Chemistry, 50.11, 1465-1473 (1978)

Questions

1–2 of 2 Questions  
  1. When it means 1/8 fitting, does it mean I can connect it with a tube with internal diameter of 1/8 or the external diameter has to be 1/8?

    1 answer
    1. This is a Swagelok fitting and 1/8" refers to the external (outside) diameter of the copper or stainless steel tubing. Linked below is a connector that can be used. Please contact your local technical service group if you need more information on how to connect Swagelok fittings.
      https://www.sigmaaldrich.com/product/supelco/22014

      Helpful?

  2. Does the product include an indicator for the consumption of sodium oxides?

    1 answer
    1. Upon the absorption of CO2, the greenish-brown material turns white due to the formation of sodium carbonate. It is designed to remove CO2 from gases used in various applications, such as carbon-hydrogen determinations, analyses of respiratory gases, and especially in quantitative organic micro-analyses. The sodium hydroxide non-fibrous silicate trapping medium is widely recognized for its high absorptive capacity and indicating properties. Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service