Skip to Content
Merck
All Photos(1)

Key Documents

503118

Supelco

Molecular Sieve 5A Moisture Trap

S-trap, 1/8 in. fitting

Synonym(s):

S-TRAP, MOLECULAR SIEVE 5A, 1/8IN FITTINGS

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$392.00

$392.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$392.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32110204

$392.00


Available to ship on16 April 2025Details


Request a Bulk Order

fitting

1/8 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Sieve 5A efficiently removes moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content.

Dimensions:
  • 200 cc Trap: 26 1/4 in. (67 cm) long × 1 in. (2.5 cm) diameter
  • 750 cc Trap: 18 in. (45.7 cm) long × 2 3/8 in. (6 cm) diameter
  • S-Trap: 7 1/2 in. (19 cm) long x 5 9/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length: 19 3/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service