Skip to Content
Merck
All Photos(1)

Key Documents

503223

Supelco

Carbon Dioxide Trap Refill

pkg of 500 cc

Sign Into View Organizational & Contract Pricing

Select a Size

500 CC
$322.00

$322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 CC
$322.00

About This Item

EC Number:
UNSPSC Code:
12000000

$322.00


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 500 cc

General description

Sodium hydroxide nonfibrous silicate material.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service