Skip to Content
Merck
All Photos(1)

Key Documents

502626

Upchurch Microtight® Tubing Sleeves

I.D. 0.015 in. (380 μm), green

Sign Into View Organizational & Contract Pricing

Select a Size

10 PKG
$55.20

$55.20


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
10 PKG
$55.20

About This Item

UNSPSC Code:
41115700

$55.20


Estimated to ship on18 April 2025


Request a Bulk Order

material

green

packaging

pkg of 10 ea

I.D.

0.015 in. (380 μm)

Looking for similar products? Visit Product Comparison Guide

General description

Use these PEEK sleeves with all Microtight fittings, to connect capillary tubing. All are 0.025 in. O.D.; normally, choose an I.D. that is 0.002-0.003 in. larger than the O.D. of the capillary tubing. Color-coded.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Microtight is a registered trademark of IDEX Health & Science, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service