Skip to Content
Merck
All Photos(1)

Key Documents

48800

Supelco

4-Bromofluorobenzene solution

certified reference material, 25,000 μg/mL in methanol

Synonym(s):

1-Bromo-4-fluorobenzene solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrF
CAS Number:
Molecular Weight:
175.00
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 624,8240,8260

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type tuning solution

packaging

ampule of 1 mL

concentration

25,000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

Fc1ccc(cc1)Br

InChI

1S/C6H4BrF/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

AITNMTXHTIIIBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

59.0 °F - open cup

Flash Point(C)

15 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John L Daristotle et al.
Acta biomaterialia, 90, 205-216 (2019-04-08)
Commercially available surgical sealants for internal use either lack sufficient adhesion or produce cytotoxicity. This work describes a surgical sealant based on a polymer blend of poly(lactic-co-glycolic acid) (PLGA) and poly(ethylene glycol) (PEG) that increases wet tissue adherence by incorporation
Nan Qin et al.
Scientific reports, 10(1), 11569-11569 (2020-07-16)
The recent emergence of new fentanyl analogues and synthetic opioids on the drug market poses a global public health threat. However, these compounds cannot typically be identified using existing analytical methods. In this study, we aimed to develop and validate
Nan Qin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1124, 82-99 (2019-06-10)
The abuse of fentanyl and its analogues, which are potent, short-acting, synthetic narcotic analgesics, has become a major issue. Many cases containing fentanyl-analogue and novel synthetic opioids have also been reported. Hence, we report the determination of fentanyl, fentanyl-analogue and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service