![4-Bromofluorobenzene solution certified reference material, 25,000 μg/mL in methanol](/deepweb/assets/sigmaaldrich/product/images/205/371/6d5fb9e0-dcee-4bc9-a944-c65fb00f6ecc/640/6d5fb9e0-dcee-4bc9-a944-c65fb00f6ecc.jpg)
4-Bromofluorobenzene solution
![4-Bromofluorobenzene solution certified reference material, 25,000 μg/mL in methanol](/deepweb/assets/sigmaaldrich/product/images/205/371/6d5fb9e0-dcee-4bc9-a944-c65fb00f6ecc/640/6d5fb9e0-dcee-4bc9-a944-c65fb00f6ecc.jpg)
Synonym(s):
1-Bromo-4-fluorobenzene solution
Empirical Formula (Hill Notation):
C6H4BrF
CAS Number:
Molecular Weight:
175.00
Products
Brand
Formula Weight
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2