Skip to Content
Merck
All Photos(1)

Key Documents

46838-U

Supelco

EPA 8260 Internal Standards Mix 1

certified reference material, 2500 μg/mL in methanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$67.50

$67.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$67.50

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

$67.50


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 8260
610

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

2500 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

General description

EPA 8260 Internal Standards Mix 1 is a multi-component solution containing chlorobenzene-d5, 1,4-dichlorobenzene-d4, and fluorobenzene as certified reference materials (CRMs).
US EPA 8000 Series methods are analytical methods for identifying and quantifying halogenated and aromatic volatile organic compounds on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites.

Application

EPA 8260 Internal Standards Mix 1 may be used as an internal standard to quantify volatile organic carbon (VOC) analytes in polyethylene pipe (PE) based samples by gas chromatography (GC) in conjunction with ion-trap mass spectrometry (ITMS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Chlorobenzene-d5
    1,4-Dichlorobenzene-d4
    Fluorobenzene

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical analysis applied to the development of a model for the determination of quality of PE pipes
Pinelli F, et al.
Journal of Analytical and Applied Pyrolysis, 89(1), 44-50 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service