Skip to Content
Merck
All Photos(1)

Key Documents

29591-U

CGA-350 Gas Cylinder Pressure Regulator

2-stage ultra high purity, parameter 0-100 psi

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,320.00

$1,320.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
$1,320.00

About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

$1,320.00


Available to ship on07 April 2025Details


Request a Bulk Order

parameter

0-100 psi

Looking for similar products? Visit Product Comparison Guide

General description

For use with hydrogen, methane, or 5% methane in argon gas cylinders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service