Skip to Content
Merck
All Photos(2)

Key Documents

29539-U

Supelco

Tenax® TA Thermal Desorption Tube

greener alternative

glass, O.D. x I.D. x L 1/4 in. × 3 1/2 in., fritted, preconditioned, pkg of 10 ea

Synonym(s):

Tenax® TA

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$1,730.00

$1,730.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$1,730.00

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

$1,730.00


Check Cart for Availability

Request a Bulk Order

Product Name

Tenax® TA, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, 60/80 mesh, preconditioned, pkg of 10 ea

material

glass TD tube

Quality Level

Agency

EPA TO-1,IP-1B

description

Sealed with TDS3 Storage Container

feature

fritted
preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax TA

particle size

60/80 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Sorbent material characterization using in-tube extraction needles as inverse gas chromatography column.: Discusses innovative applications of Tenax® TA in sorbent material characterization, providing insights into the physicochemical properties of adsorbents (Barajas XLO et al., 2017 doi:10.1002/jssc.201601198).

  • Online monitoring of trace chlorinated benzenes in flue gas of municipal solid waste incinerator by windowless VUV lamp single photon ionization TOFMS coupled with automatic enrichment system.: Details a sophisticated system integrating Tenax® TA for continuous monitoring of hazardous air pollutants in industrial settings, crucial for environmental safety and compliance (Liu et al., 2016 doi:10.1016/j.talanta.2016.09.026).

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the weight of the adsorbent bed for Tenax® TA (29539-U)?

    1 answer
    1. The weight of the adsorbent bed in 29539-U is 0.186 grams.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service