Skip to Content
Merck
All Photos(4)

Documents

29530-U

Supelco

Tenax® TA Thermal Desorption Tube

greener alternative

glass, O.D. x I.D. x L 1/4 in. × 3 1/2 in., fritted, preconditioned, pkg of 10 ea

Synonym(s):

Tenax TA Thermal Desorption Tube (glass)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Tenax® TA, 35/60 mesh, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

material

glass TD tube

Agency

EPA TO-1,IP-1B

description

Sealed with TDS³ Storage Container

feature

fritted
preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax TA porous polymer

particle size

35/60 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Determination of 67 volatile organic compounds in ambient air using thermal desorption-gas chromatography-mass spectrometry.: This research outlines the effectiveness of Tenax® TA in the broad-spectrum analysis of air quality, crucial for environmental monitoring and regulatory compliance (Zhu et al., 2019 doi:10.3724/SP.J.1123.2019.04043).

  • Chromatographic analysis of the traces of 2-chlorobenzalmalononitrile with passive adsorption from the headspace on Tenax TA and Carbotrap 300.: This study highlights the utility of Tenax® TA in forensic science for capturing and analyzing trace chemical residues, which is crucial for legal and investigative purposes (Borusiewicz, 2019 doi:10.1016/j.forsciint.2019.109933).

  • Recovery and reactivity of polycyclic aromatic hydrocarbons collected on selected sorbent tubes and analyzed by thermal desorption-gas chromatography/mass spectrometry.: This publication demonstrates the role of Tenax® TA in the efficient capture and analysis of complex organic pollutants, important for studies on pollution and its effects on health and the environment (Wallace et al., 2019 doi:10.1016/j.chroma.2019.05.030).

  • A New Method for Workplace Monitoring of Airborne Diacetyl and 2,3-Pentanedione Using Thermal Desorption Tubes and Gas Chromatography-Mass Spectrometry.: This research describes a new method utilizing Tenax® TA for monitoring harmful volatile compounds in workplace environments, contributing to occupational safety and health standards (Pengelly and Brown, 2019 doi:10.1093/annweh/wxz014).

  • Innovative coupling of supercritical fluid extraction with ion mobility spectrometry.: Outlines a novel analytical approach integrating Tenax® TA, enhancing the efficiency and sensitivity of chemical analyses, particularly useful in complex mixture separations in research and industrial applications (Jurado-Campos et al., 2018 doi:10.1016/j.talanta.2018.06.045).

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service