Skip to Content
Merck
All Photos(1)

Documents

27213

Supelco

Vials, screw top with polypropylene open-top cap, pre-assembled, pkg of 100

volume 40 mL, clear glass vial, O.D. × H 29 mm × 82 mm, tan PTFE/silicone septum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

black polypropylene cap (open-top)
clear glass vial
tan PTFE/silicone septum

feature

closure type black polypropylene hole cap
closure type screw top vial

packaging

pkg of 100 ea

O.D. × H

29 mm × 82 mm

volume

40 mL

fitting

thread for 24-400

Looking for similar products? Visit Product Comparison Guide

Application

Vials may be used in HPLC analysis, for quantifying adsorption of the three cationic membrane-active peptides mastoparan X, melittin, and magainin 2.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of cationic peptides to solid surfaces of glass and plastic.
Kristensen, Kasper, Jonas R. Henriksen, and Thomas L. Andresen.
PLoS ONE, 10.5, e0122419-e0122419 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service