Key Documents
22138-U
On/Off Throttling Valve
brass, 1/8 in.
Select a Size
1 EA
$180.00
$180.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$180.00
About This Item
UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54
$180.00
Check Cart for Availability
Request a Bulk Order
material
brass
fitting
1/8 in.
General description
Nuts and ferrules included.
Throttling valve′s on and off settings determines whether or not fluid flows past the valve. It is much more efficient than the toggle on/off valve, as it provides better flow control between 0 and 100% flow and has a larger baseline pressure drop. There are many styles of throttling valve like globe valve and butterfly valve.[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Process Plant Equipment: Operation, Control, and Reliability.
Holloway MD
Technology & Engineering, 401-402 (2012)