Skip to Content
Merck
All Photos(1)

Key Documents

22036

Swagelok® Tee

Swagelok® 400-3, stainless steel, 1/4 in. Swagelok

Synonym(s):

Swagelok® 400-3,SS TEE 1/4" EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$110.00

$110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$110.00

About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

$110.00


Check Cart for Availability

Request a Bulk Order

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 400-3

fitting

1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Two Swagelok® tees connected in series maybe used in the nanoliter injection system used for microliter column chromatography, the branch of the first tee could be connected to a restricted capillary followed by high-pressure shut-off valve (“SPLIT”), whereas the branch of the second tee could be connected with wide-bore stainless steel tubing directly to a shut-off valve (“PURGE”).[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maëlle Lorvellec et al.
iScience, 23(12), 101808-101808 (2020-12-12)
Explosion of gene therapy approaches for treating rare monogenic and common liver disorders created an urgent need for disease models able to replicate human liver cellular environment. Available models lack 3D liver structure or are unable to survive in long-term
Nanoliter injection system for microcolumn liquid chromatography.
McGuffin VL and Novotny M.
Analytical Chemistry, 55 (3), 580-583 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service