Key Documents
22013
Swagelok® Back Ferrule
Swagelok® 204-1, brass, 1/8 in. Swagelok
Synonym(s):
Swagelok® 204-1,BRASS BK FERR 1/8" PK20
Select a Size
20 EA
$41.40
$41.40
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
20 EA
$41.40
About This Item
UNSPSC Code:
41115717
$41.40
Check Cart for Availability
Request a Bulk Order
material
brass
packaging
pkg of 20 ea
manufacturer/tradename
Swagelok® 204-1
fitting
1/8 in. Swagelok
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut.[1] Glass chromatographic column require brass back ferrule, PTFE front ferrule and brass nuts for adequate sealing.[2]
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Walker JQ
Chromatographic Systems: Maintenance and Troubleshooting, 343-344 (2012)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)