Skip to Content
Merck
All Photos(1)

Key Documents

21150AST

Supelco

Supelguard HPLC Guard Cartridge Holder

Stand-Alone (Swivel-type), for use with Supelguard cartridges (2 cm L. x 2 to 4.6 mm I.D.)

Synonym(s):

Guard Cartridge System

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$444.00

$444.00


Check Cart for Availability


Select a Size

Change View
1 EA
$444.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$444.00


Check Cart for Availability

Product Name

Supelguard Guard Cartridge Holder, Stand-Alone (Swivel-type), for use with Supelguard cartridges (2 cm L. x 2 to 4.6 mm I.D.)

material

stainless steel holder

Quality Level

description

swivel design

packaging

pkg of 1 ea

fitting

10-32 female (Valco compatible)

compatibility

for use with Supelguard cartridges (2 cm L. x 2 to 4.6 mm I.D.)

Looking for similar products? Visit Product Comparison Guide

General description

Stand-alone guard column holder fits 2 cm x 2, 3, and 4 mm I.D. Astec® Cellulose DMP Chiral HPLC guard cartridges. It accommodates standard 1/16" O.D. Valco-type nuts and ferrules. One end of this holder has a freely rotating inlet/outlet port, which allows for complete rotation of tubing on one side of the holder. Cartridges, tubing, nuts, and ferrules are not included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

In this study, optimized methods are presented for sample preparation and chiral chromatography for the LC/MS analysis of amphetamine and methamphetamine enantiomers in urine.

Related Content

CYCLOBOND is the name given to Astec technology for bonding cyclodextrins to a high purity silica gel through a stable ether linkage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service