Accéder au contenu
Merck
HomeLarge Molecule HPLCAstec CYCLOBOND
Astec CYCLOBOND
CYCLODEXTRIN Stationary Phases for Chiral Separations and Highly Selective Achiral Separations
CYCLOBOND is the name given to Astec technology for bonding cyclodextrins to a high purity silica gel through a stable ether linkage. Introduced in 1983, this patented stationary phase retains ability to form inclusion complexes, and allows for numerous chemical separations by selectively including into the cyclodextrin cavity, from solution, a wide variety of organic molecules. CYCLOBOND I are β-cyclodextrin bonded and CYCLOBOND II are γ-cyclodextrin bonded.
CYCLOBOND I 2000 series of HPLC columns is specially formulated to meet today’s stringent requirements for analysis in the pharmaceutical industry, and for small analytes of general interest in the chemical and environmental areas. Astec has focused on the need to accurately and reproducibly separate enantiomers. The result is a high performance range of chiral separation phases with guaranteed batch to batch reproducibility, greater stability and improved selectivity and resolution. Based on the original CYCLOBOND I (β-cyclodextrin) columns, the CYCLOBOND I 2000 series are second-generation products. The native β-cyclodextrin and eight β-cyclodextrin derivatives are in the CYCLOBOND I 2000 series.
CYCLOBOND II series columns are excellent chiral selectors for multi-ring structures such as those based on anthracene, chrysene or pyrene. These are γ-cyclodextrin bonded phases, and consist of 8 glucopyranose units arranged in the same truncated cone shape. Applications include steroids, porphorins, FMOC amino acids. Applications for CYCLOBOND II AC, an acetylated derivative, include napthylethylamine, steroids, and sterols.
Ordering Information
CYCLOBOND I 2000
Loading
CYCLOBOND I 2000 AC
Loading
CYCLOBOND I 2000 SP
Loading
CYCLOBOND I 2000 RSP
Loading
CYCLOBOND I 2000 HP-RSP
Loading
CYCLOBOND I 2000 DMP
Loading
CYCLOBOND I 2000 DM
Loading
CYCLOBOND I 2000 DNP
Loading
CYCLOBOND II
Loading
CYCLOBOND II AC
Loading
Connectez-vous pour continuer
Pour continuer à lire, veuillez vous connecter à votre compte ou en créer un.
Vous n'avez pas de compte ?