Skip to Content
Merck
All Photos(1)

Key Documents

567278-U

Supelco

Discovery® BIO Wide Pore C5 (3 µm) HPLC Columns

L × I.D. 2 cm × 2.1 mm, pkg of 1 Kit

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$444.00

$444.00


Check Cart for Availability


Select a Size

Change View
2 EA
$444.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$444.00


Check Cart for Availability

Product Name

Discovery® BIO Wide Pore C5 Supelguard Cartridge, 3 μm particle size, L × I.D. 2 cm × 2.1 mm

material

stainless steel column

Quality Level

product line

Discovery®

feature

endcapped

packaging

pkg of 2 ea

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

2 cm × 2.1 mm

surface area

100 m2/g

matrix

fully porous particle

matrix active group

C5 (pentyl) phase

particle size

3 μm

pore size

300 Å

operating pH

2-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Kits include one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules. Guard cartridges require holders that are sold separately. The 2.1 and 4 mm I.D. cartridges use 21150AST or 59660-U (both stand-alone) or 504254 or 55205 (both integral). The 10 mm I.D. cartridges use 567499-U. The 21.2 mm I.D. cartridges use 581392-U.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discovery BIO Wide Pore C5 products offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service