Skip to Content
Merck
All Photos(1)

Key Documents

Z740336

ThermASeal Foil heat sealable seal

VII LabRoll, W × L 115 mm × 350 m, sterile

Synonym(s):

bonding foil, non pierable bonding foil

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,660.00

$2,660.00


Check Cart for Availability


Select a Size

Change View
1 EA
$2,660.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

$2,660.00


Check Cart for Availability

sterility

sterile

feature

piercable: no

packaging

pack of 1 ea

manufacturer/tradename

IST Scientific IST-108-115SR

W × L

115 mm × 350 m

Looking for similar products? Visit Product Comparison Guide

General description

Strong bonding foil to polypropylene, ideal for water thermal cyclers.

Features: Foil laminate, strong bonding, good solvent resistance including DMSO, peelable, non-pierceable.
Compatibility: Polypropylene (PP), Polystyrene (PS).
Seal integrity range: -200°C to 110°C.

Application

PCR, Ideal for water bath thermo cyclers, long term compound storage for hazardous materials

Legal Information

LabRoll is a trademark of IST Scientific
ThermASeal Foil is a trademark of IST Scientific

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service