Skip to Content
Merck
All Photos(1)

Key Documents

CLS3358

Corning® 96 Well Clear Polystyrene Microplate

round bottom clear, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$818.00

$818.00


Available to ship on07 April 2025Details



Select a Size

Change View
100 EA
$818.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
clear polystyrene
round bottom clear
round wells
size:
96 wells
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
skirt

$818.00


Available to ship on07 April 2025Details


material

clear polystyrene
round bottom clear
round wells

sterility

sterile

feature

lid
skirt

packaging

bag of 20

manufacturer/tradename

Corning 3358

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Wells with rounded bottoms having individual α-numeric codes for identification
  • Plates are treated for optimal cell attachment
  • Nonpyrogenic in nature and gamma radiation sterilized
  • Surface is negatively charged and is hydrophilic in nature
  • Free from animal-derived materials

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service