CLS6570
Corning® microplate sealing tape
aluminum film, non-sterile, suitable for (for short and long term storage and PCR, 96 well reactions)
Synonym(s):
microplate sealing film, multiwell plate sealing film, sealing film
Select a Size
$92.60
Available to ship on17 April 2025Details
Select a Size
About This Item
$92.60
Available to ship on17 April 2025Details
Recommended Products
material
aluminum film
sterility
non-sterile
packaging
case of 100 ea
manufacturer/tradename
Corning 6570
technique(s)
PCR: suitable
size
96 wells
suitability
suitable for (for short and long term storage and PCR, 96 well reactions)
application(s)
agriculture
Looking for similar products? Visit Product Comparison Guide
General description
- Easy application and removal for short- and long-term storage
- Provide tight seal to minimize evaporation and condensation and enables oil-free operation when used with PCR thermocyclers with heated lids
- Suitable for use between -80°C and 150°C, is not transparent, and is pierceable
- DMSO-resistant
- Nonsterile
- Certified DNase- and RNase-free
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service