Skip to Content
Merck
All Photos(1)

Documents

Z722995

Nunc® 384 well polystyrene plates

384 well plate, non-treated, polystyrene, 120ul/well, pinchbar design, 100/cs

Synonym(s):

384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

black polystyrene
flat bottom black wells

sterility

non-sterile

feature

lid: no

packaging

case of 100 ea (internal packs of 25)

manufacturer/tradename

Nunc 262260

plate L × W

128 mm × 86 mm

size

384 wells

well maximum volume

100 μm

working volume

100 μL

color

black

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Nunc® 384 Well Plates, Polystyrene

  • Cell culture surface available (Nunclon® Δ)
  • Non-treated surface for storage, assays and screening
  • MaxiSorp and PolySorp® for immunoassays; ELISA
  • Clear plates for colorimetric assays and sample storage
  • White plates for luminescence and fluorescence assays
  • Black plates for fluorescence assays
  • White and black plates minimize crosstalk
  • Rounded square wells eliminate wicking (capillary action)
  • Flat bottom for optical reading
  • Optimized for scintillation counting
  • Working volume range: 10-100 μl/well

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service