Skip to Content
Merck
All Photos(1)

Key Documents

Z723010

Nunc® 384 well polystyrene plates

size 384 wells, non-treated surface, flat bottom clear polystyrene wells, maximum volume 120 μL

Synonym(s):

384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$1,190.00

$1,190.00


Check Cart for Availability


Select a Size

Change View
100 EA
$1,190.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear polystyrene wells
polystyrene
size:
384 wells
sterility:
non-sterile
binding type:
non-treated surface
feature:
lid: no

$1,190.00


Check Cart for Availability

material

clear polystyrene
flat bottom clear polystyrene wells
polystyrene

sterility

non-sterile

feature

lid: no

packaging

case of 100 ea (internal packs of 25)

manufacturer/tradename

Nunc 262160

maximum volume

120 μL

plate L × W

128 mm × 86 mm

size

384 wells

working volume

100 μL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Nunc® 384 Well Plates, Polystyrene

  • Cell culture surface available (Nunclon® Δ)
  • Non-treated surface for storage, assays and screening
  • MaxiSorp and PolySorp® for immunoassays; ELISA
  • Clear plates for colorimetric assays and sample storage
  • White plates for luminescence and fluorescence assays
  • Black plates for fluorescence assays
  • White and black plates minimize crosstalk
  • Rounded square wells eliminate wicking (capillary action)
  • Flat bottom for optical reading
  • Optimized for scintillation counting
  • Working volume range: 10-100 μl/well

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service