Skip to Content
Merck
All Photos(1)

Key Documents

Z359076

Nalgene® Labtop cooler

0°C Labtop cooler, Jr., transparent lid

Synonym(s):

PCR amplification cooler, PCR amplification workstation

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$511.00

$511.00


Check Cart for Availability


Select a Size

Change View
1 EA
$511.00

About This Item

UNSPSC Code:
41103025
NACRES:
NB.22

$511.00


Check Cart for Availability

feature

Holds cryognic vials and 0.2 to 0.5mL microcentrifuge tubes

manufacturer/tradename

Nalgene® DS5116-0012

L × W × H

151 mm × 108 mm × 125 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Maintains 12 microtubes below 1°C for up to 3 1/2 hrs at room temperature.
Molded from durable polycarbonate and filled with non-toxic freezable gel. A convenient alternative to ice buckets for keeping reagents and enzymes cool. Also protects critical samples from temperature fluctuations in the freezer.

Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service