Documents
Z370509
Nalgene® Labtop cooler
0°C Labtop cooler, clear lid
Synonym(s):
PCR amplification cooler, PCR amplification workstation
About This Item
UNSPSC Code:
41103025
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 5116-1300
L × W × H
197 mm × 140 mm × 190 mm
Related Categories
General description
Maintains twelve 12-13 mm diameter tubes below 1°C for up to 5 hrs at room temperature.
Molded from durable polycarbonate and filled with non-toxic freezable gel. A convenient alternative to ice buckets for keeping reagents and enzymes cool. Also protects critical samples from temperature fluctuations in the freezer.
Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries