Skip to Content
Merck
All Photos(1)

Key Documents

Z0502

Sigma-Aldrich

Zaleplon

≥98% (HPLC), solid

Synonym(s):

CL-284846, N-[3-(3-Cyanopyrazolo-[1,5-a]pyrimidin-7-yl)phenyl)-N-ethylacetamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H15N5O
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

drug control

USDEA Schedule IV; psychotrope (France); regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated
protect from light

color

white

solubility

DMSO: ~20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CCN(C(C)=O)c1cccc(c1)-c2ccnc3c(cnn23)C#N

InChI

1S/C17H15N5O/c1-3-21(12(2)23)15-6-4-5-13(9-15)16-7-8-19-17-14(10-18)11-20-22(16)17/h4-9,11H,3H2,1-2H3

InChI key

HUNXMJYCHXQEGX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Zaleplon is a non-benzodiazepine agonist at the benzodiazepine site of the GABAA receptor; sedative; hypnotic.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monique A J Mets et al.
Sleep medicine reviews, 14(4), 259-267 (2010-02-23)
Disturbed body balance and standing steadiness are problematic for those who wake up at night or in the morning after using hypnotic drugs. As a result, falls and hip fractures are frequently reported in patients using sleep medication. A literature
Daniela Remane et al.
Analytical and bioanalytical chemistry, 401(4), 1341-1352 (2011-07-21)
For fast and reliable screening, identification, and quantification of as many analytes as possible, multi-analyte approaches are very useful in clinical and forensic toxicology. Using ultra high performance liquid chromatography-tandem mass spectrometry, such an approach has been developed for blood
K W Weitzel et al.
Clinical therapeutics, 22(11), 1254-1267 (2000-12-16)
Insomnia is the subjective complaint of poor sleep or an inadequate amount of sleep that adversely affects daily functioning. For the past 4 decades, treatment of insomnia has shifted away from the use of barbiturates toward the use of hypnotic
M Dooley et al.
Drugs, 60(2), 413-445 (2000-09-13)
Zaleplon is a pyrazolopyrimidine hypnotic agent which is indicated for the short term (2 to 4 weeks) management of insomnia. Zaleplon 5 and 10 mg at bedtime (usual recommended doses) significantly reduced sleep latency compared with placebo in clinical trials
Ane Birgitte Telén Andersen et al.
Pharmacoepidemiology and drug safety, 20(4), 378-385 (2011-01-25)
To estimate the degree of long-term use of zopiclone, zolpidem and zaleplon among Danes aged 65 and older and the association with sociodemographic factors and use of other drugs. Danish register-based study of 5000 men and 5000 women aged 65

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service