Zaleplon
Synonym(s):
CL-284846, N-[3-(3-Cyanopyrazolo-[1,5-a]pyrimidin-7-yl)phenyl)-N-ethylacetamide
Empirical Formula (Hill Notation):
C17H15N5O
CAS Number:
Molecular Weight:
305.33
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing