Skip to Content
Merck
All Photos(1)

Key Documents

V3877

Sigma-Aldrich

4-Vinylpyridine

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N
CAS Number:
Molecular Weight:
105.14
Beilstein:
104506
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

contains

100 ppm hydroquinone

refractive index

n20/D 1.549 (lit.)

bp

62-65 °C/15 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C=Cc1ccncc1

InChI

1S/C7H7N/c1-2-7-3-5-8-6-4-7/h2-6H,1H2

InChI key

KFDVPJUYSDEJTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the modification of cysteine.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino acid analysis.
J Ozols
Methods in enzymology, 182, 587-601 (1990-01-01)
Single hydrolysis method for all amino acids, including cysteine and tryptophan.
A S Inglis
Methods in enzymology, 91, 26-36 (1983-01-01)
Jong Hyun Kim et al.
ACS nano, 10(3), 3342-3355 (2016-02-24)
Although previous studies suggest that nanotopographical features influence properties and behaviors of stem cells, only a few studies have attempted to derive clinically useful somatic cells from human pluripotent stem cells using nanopatterned surfaces. In the present study, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service