Skip to Content
Merck
All Photos(4)

Key Documents

V2909

Sigma-Aldrich

2-Vinylnaphthalene

greener alternative

95%

Synonym(s):

β-Vinylnaphthalene, 2-Ethenylnaphthalene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$207.00
25 G
$761.00

$207.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$207.00
25 G
$761.00

About This Item

Linear Formula:
C10H7CH=CH2
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$207.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

greener alternative product score

old score: 49
new score: 26
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

<5% methyl alcohol

mp

64-68 °C (lit.)

greener alternative category

storage temp.

2-8°C

SMILES string

C=Cc1ccc2ccccc2c1

InChI

1S/C12H10/c1-2-10-7-8-11-5-3-4-6-12(11)9-10/h2-9H,1H2

InChI key

KXYAVSFOJVUIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Vinylnaphthalene (2VNP) can be prepared by the alkylation of naphthalene to I-and 2 ethylnapthalenes followed by dehydrogenation, or by oxidation followed by dehydration. 2VNP is the monomer for poly-(2-vinylnapthalene).[1]
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Application

2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer[2] and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene.The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.[3]

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT SE 2

Target Organs

Eyes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies of the antenna effect in polymer molecules. 13 preparation and photophysical studies of poly (sodium styrenesulfonate-co-2-vinylnaphthalene).
Nowakowska MB, et al.
Macromolecules, 22(10), 3903-3908 (1989)
Alger MSM
Polymer Science Dctionary (1989)
Poly (2-vinylnaphthalene-alt-maleic acid)-graft-polystyrene as a Photoactive Polymer Micelle and Stabilizer for Polystyrene Latexes
Cao T, et al.
Macromolecules, 27(25), 7459-7464 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service