Skip to Content
Merck
All Photos(3)

Key Documents

335681

Sigma-Aldrich

Ethylene glycol dimethacrylate

98%, cross-linking reagent polymerization reactions, methacrylate, 90-110 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Polyethylene glycol, 1,2-Ethanediol dimethacrylate, Ethylene dimethacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$79.80
100 ML
$103.00
500 ML
$285.00

$79.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$79.80
100 ML
$103.00
500 ML
$285.00

About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH2OCOC(CH3)=CH2
CAS Number:
Molecular Weight:
198.22
Beilstein:
1776663
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$79.80


Check Cart for Availability

Request a Bulk Order

Product Name

Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor

vapor density

>1 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 21.1 °C)

Assay

98%

form

liquid

contains

90-110 ppm monomethyl ether hydroquinone as inhibitor

reaction suitability

reagent type: cross-linking reagent
reaction type: Polymerization Reactions

refractive index

n20/D 1.454 (lit.)

bp

98-100 °C/5 mmHg (lit.)

density

1.051 g/mL at 25 °C (lit.)

Ω-end

methacrylate

α-end

methacrylate

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCCOC(=O)C(C)=C

InChI

1S/C10H14O4/c1-7(2)9(11)13-5-6-14-10(12)8(3)4/h1,3,5-6H2,2,4H3

InChI key

STVZJERGLQHEKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol dimethylacrylate (EGDMA) is a diester formed by condensation of two equivalents of methacrylic acid and one equivalent of ethylene glycol.

Application

Ethylene glycol dimethacrylate can be used as a cross-linker in thepreparation of:

  • A terpolymer system, poly(acrylonitrile-co-Ethyleneglycol dimethacrylate-co-vinylbenzyl chloride), which is applicable as a sorbentin pharmaceuticals adsorption.
  • PVA (polyvinyl alcohol) based hydrogel by free-radicalcopolymerization. These PVA polymer networks can be used as drug deliverysystems.
It can also be used asa monomer in the preparation of swellable and non-swellable poly(ethyleneglycol dimethacrylate/acrylic acid) copolymer microspheres through suspensionpolymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - Pensky-Martens closed cup

Flash Point(C)

104 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Qi et al.
ACS applied materials & interfaces, 11(32), 28690-28698 (2019-07-20)
Compared with conventional glass slides and two-dimensional (2D) planar microarrays, polymer-based support materials and three-dimensional (3D) surface structures have attracted increasing attention in the field of biochips because of their good processability in microfabrication and low cost in mass production
Kouroush Salimi et al.
Journal of chromatography. A, 1496, 9-19 (2017-03-30)
A seeded polymerization protocol was developed for the synthesis of monodisperse-porous poly(vinylphosphonic acid-co-ethylene dimethacrylate), [poly(VPA-co-EDMA)] microspheres with superior porous properties. The protocol allowed the direct synthesis of phosphonic acid functionalized porous microspheres with the mean size of ∼4μm and the
Zhen Wang et al.
Electrophoresis, 33(21), 3151-3158 (2012-09-06)
A microfluidic device that performs "in space" sample fractionation, collection, and preconcentration for proteomics is described. Effluents from a 2.75 mm long fractionation channel, focused via sheath flow, were sequentially delivered into an array of 36-collection channels containing monolithic polymer
Mariane Gonçalves Santos et al.
Talanta, 163, 8-16 (2016-11-26)
The use of a new class of hybrid materials, called restricted access molecularly imprinted polymers (RAMIPs) seems to present a good strategy for the sample preparation of complex matrices, since these materials combine good protein elimination capacity with high degree
Andreia F R Pimenta et al.
PloS one, 11(12), e0167728-e0167728 (2016-12-10)
The study of ocular drug delivery systems has been one of the most covered topics in drug delivery research. One potential drug carrier solution is the use of materials that are already commercially available in ophthalmic lenses for the correction

Articles

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Is the ethylene glycol dimethacrylate a toxic crosslinker for human?

    1 answer
    1. The toxicological information for product 335681 can be reviewed in the SDS.
      See the link below, section 13, to review this information:
      https://www.sigmaaldrich.com/sds/aldrich/335681

      There are articles available in the literature as well. See the link to the following publication for an example:
      https://pubmed.ncbi.nlm.nih.gov/29107684/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service