Skip to Content
Merck
All Photos(1)

Key Documents

SRE0062

Sigma-Aldrich

Tris-Borate EDTA Buffer, 10X Concentrate

Synonym(s):

TBE Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$141.00
4 L
$327.00

$141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$141.00
4 L
$327.00

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25

$141.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

application(s)

diagnostic assay manufacturing

storage temp.

room temp

Application

Tris-Borate EDTA (TBE) buffer is commonly used in the electrophoresis of nucleic acids in agarose and polyacrylamide gels.

TBE buffer is recommended for resolution of RNA and DNA fragments smaller than 1500 bp.

Physical form

10X TBE buffer stock solution contains 0.89 M Trizma(R) with 0.02 M EDTA adjusted to pH 8.3 with boric acid. Dilute with ultrapure water to 1X.

Notes: If precipitation is present in the 10X concentrated buffer, warm the bottle to 37 °C and mix until completely dissolved prior to dilution.
It is recommended 1X working solutions be filtered through a 0.2 mm filter before use.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service