Skip to Content
Merck
All Photos(1)

Key Documents

93290

Sigma-Aldrich

TRIS borate – EDTA buffer solution

for Northern and Southern blotting, solution

Synonym(s):

TBE buffer

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$59.00
5 L
$284.00

$59.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 L
$59.00
5 L
$284.00

About This Item

UNSPSC Code:
41105319
eCl@ss:
32129211
NACRES:
NA.25

$59.00


Estimated to ship on16 April 2025


Request a Bulk Order

grade

for molecular biology

Quality Level

description

long run, concentrate

product line

BioReagent

form

solution

quality

10× concentrate
ready-to-use solution

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

8.6-9.0

storage temp.

room temp

General description

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. The TBE solution is effective under slightly basic conditions, which keeps DNA deprotonated , water-soluble, and protected from degradation. This concentrate can be easily diluted to 1x or 0.5x before use (with molecular biology grade water).

Application

Suitable for use as running buffer for nucleic acid electrophoresis, which may be part of Northern and Southern blot hybridization procedures.

Components

The TBE 10x liquid contains 1.3M TRIS, 450 mM boric acid and 25 mM EDTA in water.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Johnston-Dow et al.
Biotechniques, 5, 754-754 (1987)

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service