574797
TAE Buffer, 10X, Molecular Biology Grade
A 10X concentrate that can be diluted to a 1X solution containing 40 mM Tris, 40 mM acetate, and 1 mM EDTA, pH ~8.3.
Synonym(s):
Tris-Acetate-EDTA Buffer
About This Item
Recommended Products
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated
technique(s)
electrophoresis: suitable
foreign activity
DNase, RNase, protease, none detected
shipped in
ambient
storage temp.
15-25°C
General description
Application
Warning
Preparation Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service