Skip to Content
Merck
All Photos(1)

Key Documents

SML3592

Sigma-Aldrich

GSK3179106

≥98% (HPLC)

Synonym(s):

2-(4-(4-ethoxy-6-oxo-1,6-dihydropyridin-3-yl)-2-fluorophenyl)-N-(5-(1,1,1-trifluoro-2-methylpropan-2-yl)isoxazol-3-yl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$194.00
25 MG
$783.00

$194.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$194.00
25 MG
$783.00

About This Item

Empirical Formula (Hill Notation):
C22H21F4N3O4
CAS Number:
Molecular Weight:
467.41
UNSPSC Code:
12352200
NACRES:
NA.21

$194.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C(C)(C)c1[o]nc(c1)NC(=O)Cc2c(cc(cc2)c3c[nH][c](cc3OCC)=O)F

InChI

1S/C22H21F4N3O4/c1-4-32-16-9-19(30)27-11-14(16)12-5-6-13(15(23)7-12)8-20(31)28-18-10-17(33-29-18)21(2,3)22(24,25)26/h5-7,9-11H,4,8H2,1-3H3,(H,27,30)(H,28,29,31)

InChI key

IDXKJSSOUXWLDB-UHFFFAOYSA-N

Biochem/physiol Actions

GSK3179106 is a potent and selective gut-restricted RET (Rearranged during transfection) kinase inhibitor. GSK3179106 attenuates abdominal contractions induced by colorectal distention in rat models.
Potent and selective gut-restricted RET (Rearranged during transfection) kinase inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploring the Potential of RET Kinase Inhibition for Irritable Bowel Syndrome: A Preclinical Investigation in Rodent Models of Colonic Hypersensitivity
Journal of Pharmacology and Experimental Therapeutics, 368, 299-307 (2019)
Discovery of a First-in-Class Gut-Restricted RET Kinase Inhibitor as a Clinical Candidate for the Treatment of IBS
ACS Medicinal Chemistry Letters, 9, 623-628 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service